During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.

CellSensor™ irf1-bla TF-1 Cell Line

Catalog number:  K1657

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Jak/Stat signaling pathways play essential roles in cellular responses to distinct cytokines. One of the Jak/Stat pathways, Jak2/Stat5, mediates cell proliferation in response to Interleukin - 3 (IL - 3), prolactin, erythropoietin (Epo), and granulocyte - macrophage colony stimulating factor (GM - CSF). JAK2 gene knockout causes embryonic lethality due to defective erythropoiesis, suggesting that the Jak2/Stat5 pathway plays an important role in red blood cell formation. The recent discovery of an activating mutation in JAK2 (V617F) present in a high percentage of myeloproliferative disease (MPD) patients suggests that the Jak2/Stat5 pathway may be a potential therapeutic target for certain forms of MPD. The activated Stat5 transcription factor recognizes and binds to a specific palindromic DNA sequence found in the promoter region of β - casein, interferon regulatory factor - 1 (irf - 1), and a number of other genes. The CellSensor® irf1 - bla TF1 Cell Line contains a beta - lactamase reporter gene under control of the irf - 1 response element stably integrated into TF1 cells. TF1 cells are a human erythroleukemia cell line that is growth - dependent on GM - CSF and have an intact GM - CSF - JAK2 - STAT5 pathway. This cell line is validated for EC50 and Z’ - factor using GM - CSF. This cell line has also been tested under variable experimental conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time. Responsiveness to EPO and Jak Inhibitor 1 was also tested (Figures 2 and 3). CellSensor® irf1 - bla TF1 cells were stimulated in triplicate with GM - CSF over the indicated concentration range in a 384 - well format. Cells were incubated for 5 hrs with agonist and 0.5% DMSO and then combined with LiveBLAzer™ - FRET B/G Substrate (CCF4 - AM) for 2.5 hrs. Fluorescence emission values at 460 nm and 530 nm were obtained using a standard fluorescence plate reader. The 460/530 ratios were plotted for each replicate against the indicated concentrations of GM - CSF. CellSensor® irf1 - bla TF1 cells were stimulated with Epo over the indicated concentration range in a 384 - well format. Cells were incubated for 5 hrs with agonist and 0.5% DMSO and then combined with LiveBLAzer™ - FRET B/G Substrate (CCF4 - AM) for 2.5 hrs. Fluorescence emission values at 460 nm and 530 nm were obtained using a standard fluorescence plate reader. The Response Ratios were plotted against the indicated concentrations of Epo. CellSensor® irf1 - bla TF1 cells were treated with Jak Inhibitor 1 over the indicated concentration range in a 384 - well format. Cells were then stimulated with GM - CSF or EPO for 5 hrs in 0.5% DMSO and then combined with LiveBLAzer™ - FRET B/G Substrate (CCF4 - AM) for 2.5 hrs. Fluorescence emission values at 460 nm and 530 nm were obtained using a standard fluorescence plate reader. These values were converted to percent inhibition relative to a set of controls (unstimulated cells and EC80 of GM - CSF or EPO treated cells) and plotted against the indicated concentrations of Jak Inhibitor 1. Academic and non-profit customers, please inquire for special pricing.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell State: Dividing Cells
Druggable Target: Cytokines, Signaling Pathway
Product Line: CellSensor™
Product Size: 1 vial
System: CellSensor™

Contents & storage

CellSensor® Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.

Documents

Manuals & protocols