During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.

GeneBLAzer™ TRα-UAS-bla HEK 293T Assay Kit

Catalog number: K1684

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

TR alpha-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Thyroid hormone receptor alpha(TR alpha) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer®UAS-bla HEK 293T cell line. GeneBLAzer®UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-TR alpha (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. TR alpha-UASbla HEK 293T 293 cells are functionally validated for Z' and EC50 concentrations of T3 Thryoid hormone. In addition, TR alpha-UAS-bla HEK 293T 293 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Alternate Agonists: 9-cis-retinoic acid, AM 580, CD437, TTNPB
Antagonists: AGN 193109
Cell State: Dividing Cells
Detection Method: Fluorescent
Druggable Target: Nuclear Receptors
Primary Agonist: All-trans-retinoic Acid
Product Line: GeneBLAzer®
Product Size: 1 plate
Reporter: BLA (Beta-Lactamase)
System: GeneBLAzer™

Contents & storage

Shipping: Dry Ice Storage: -80°C/Liquid Nitrogen Contains: TR alpha-UAS-bla HEK 293T cells (K1260)

Documents

Manuals & protocols