During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.

GeneBLAzer™ RARα-UAS-bla HEK 293T Cells

Catalog number: K1685

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

RAR alpha-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human retinoic acid receptor alpha fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer®UAS-bla HEK293T cell line. GeneBLAzer®UAS-bla HEK 293T cells (catalog#K1104) stably express a beta-lactamase reporter gene under the transcriptional control of a 7x Upstream Activator Sequence (UAS). Transcription from the 7xUAS is activated by the binding of the GAL4 transcription factor DNA-binding-domain (DBD). The GAL4-DBD is expressed as a fusion protein with the ligand binding domain (LBD) of RAR alpha. When an agonist binds to the LBD of the GAL4(DBD)-RAR alpha(LBD) fusion protein it translocates to the nucleus where it binds to the 7x UAS inducing transcription of betalactamase. RAR alpha-UAS-bla HEK 293T cells have been tested for assay performance using variable assay conditions, including DMSO concentration, cell number, stimulation time, substrate loading time and have been validated for Z' and EC50 concentrations of all-trans retinoic acid. Additional testing data using alternate stimuli are also available.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Alternate Agonists: T4, Tetrac
Antagonists: 1-850
Cell State: Dividing Cells
Detection Method: Fluorescent
Druggable Target: Nuclear Receptors
Primary Agonist: T3 Hormone
Product Line: GeneBLAzer®
Product Size: 2 x 106 cells
Reporter: BLA (Beta-Lactamase)
System: GeneBLAzer™

Contents & storage

Shipping: Dry Ice Storage: -80°C/Liquid Nitrogen Contains: TR alpha-UAS-bla HEK 293T cells (K1260)

Documents

Manuals & protocols

Recommended products