During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.

GeneBLAzer™ RARβ-UAS-bla HEK 293T Cells

Catalog number:  K1695

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


RAR beta-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human retinoic acid receptor beta (RAR beta) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer®UAS-bla HEK 293T cell line. GeneBLAzer®UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-RAR beta (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. RAR beta-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of all-trans retinoic acid (ATRA). In addition, RAR beta-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, and stimulation time.
For Research Use Only. Not for use in diagnostic procedures.


Alternate Agonists: All-trans-retinoic acid, Docosahexaenoic Acid, Phytanic Acid
Antagonists: none
Cell State: Dividing Cells
Detection Method: Fluorescent
Druggable Target: Nuclear Receptors
Primary Agonist: 9-cis-retinoic acid
Product Line: GeneBLAzer®
Product Size: 2 x 106 cells
Reporter: BLA (Beta-Lactamase)
System: GeneBLAzer™

Contents & storage

GeneBLAzer® Nuclear Receptor Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.


Manuals & protocols

Recommended products