During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.

GeneBLAzer® D2-Gqo5-NFAT-bla CHO-K1 Cells

Catalog number: K1708

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

GeneBLAzer® D2-Gqo5-NFAT-bla CHO-K1 cells contain the human Dopamine Receptor 2 (D2) stably integrated into the GeneBLAzer® Gqo5-NFAT-bla CHO-K1 cell line. GeneBLAzer® Gqo5-NFAT-bla CHO-K1 cells (K1536) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element and the Chimeric G protein, Gqo5.

The GeneBLAzer® D2-Gqo5-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of apomorphine. In addition, GeneBLAzer® D2-Gqo5-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

The D2 receptor is found primarily in the brain. There are two forms of D2 which are created by alternate splicing: a short form and a long form. This assay uses the long form, although little difference in ligand binding or signaling has been seen between the two forms.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gene Symbol: D2
Accession Number: NM_000795
Cell Line: CHO-K1
Cell State: Dividing Cells
Detection Method: Fluorescent
Druggable Target: GPCRs
Population Status: Single Cell Clone Origin
Product Line: GeneBLAzer®
Product Size: 2 x 106 cells
Reporter: BLA (Beta-Lactamase)
System: GeneBLAzer™
Technique: FRET

Contents & storage

• GeneBLAzer® D2-Gqo5-NFAT-bla CHO-K1 cells
• GeneBLAzer® D2-Gqo5-NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer® D2-Gqo5-NFAT-bla CHO-K1 cells should be stored in liquid nitrogen immediately upon receipt.

LiveBLAzer™-FRET B/G Loading Kit, Solution D, and other materials are required separately; please refer to the protocol.

Documents

Manuals & protocols