During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.

GeneBLAzer® 5TH1A-Gα15-NFAT-bla CHO-K1 Cells

Catalog number: K1712

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

GeneBLAzer® 5HT1A Gα15-NFAT-bla CHO-K1 cells contain the human 5-HT Receptor 1A (5HT1A) stably integrated into the GeneBLAzer® Gα15-NFAT-bla CHO-K1 cell line. GeneBLAzer® Gα15-NFAT-bla CHO-K1 cells (Cat. no. K1537) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element and the Promiscuous G protein, Gα15.

The GeneBLAzer® 5HT1A Gα15-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of 5-CT. In addition, GeneBLAzer® 5HT1A Gα15-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

Agonists and antagonists of the 5HT receptors are in various stages of clinical trials for the treatment of neurological disorders like anxiety, insomnia, and depression.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gene Symbol: 5HT1A
Accession Number: NM_000524
Cell Line: CHO-K1
Cell State: Dividing Cells
Detection Method: Fluorescent
Druggable Target: GPCRs
Population Status: Single Cell Clone Origin
Product Line: GeneBLAzer®
Product Size: 2 x 106 cells
Reporter: BLA (Beta-Lactamase)
System: GeneBLAzer™
Technique: FRET

Contents & storage

Contents:
K1712 GeneBLAzer® 5HT1A Gα15-NFAT-bla CHO-K1 cells
(LiveBLAzer™-FRET B⁄G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
• GeneBLAzer® 5HT1A Gα15-NFAT-bla CHO-K1 cells
• GeneBLAzer® 5HT1A Gα15-NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis

Storage:
The GeneBLAzer® 5HT1A Gα15 CHO-K1 DA cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.

Documents

Manuals & protocols