Invitrogen™

EasySelect™ Pichia Expression Kit

Catalog number:  K174001

 Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The EasySelect™ Pichia Expression Kit provides all needed components for protein production in the yeast Pichia pastoris. The EasySelect™ Kit contains Pichia yeast strains, expression vectors, and reagents that enable transformation of Pichia cells. Selection with Zeocin™ antibiotic makes it easy to screen for high-copy number transformed variants.

Advantages of the Pichia pastoris expression system:
• Culture Pichia cells as easily as E. coli
• Obtain higher cell density than other microbial expression systems.
• Generate more protein per cell than other microbial expression systems.
• Benefit from eukaryotic protein processing, protein folding, and post-translational modifications.
• Easily scale-up from shake flasks up to industrial-scale fermentation.

A Proven Protein Expression System
Over the past 30 years, Pichia pastoris has been used by labs and industry around the world for producing hundreds of different proteins from many species including human (Ref 1, 2, 3). Pichia pastoris may be grown as easily as bacteria (E. coli) or yeast (Saccharomyces cerevisiae) yet offers many advantages over those systems (Ref 4). Pichia pastoris can grow to much higher cell densities than bacteria or other yeast. Pichia pastoris is capable of growing on methanol as a sole carbon source making scale-up to industrial levels relatively inexpensive.

The Power of the AOX1 Promoter
The EasySelect™ Pichia Expression Kit uses the promoter from the Pichia alcohol oxidase 1 (AOX1) gene to drive production of your protein. Pichia pastoris is capable of growing on methanol as its only carbon source. When grown on methanol as sole carbon source, AOX1 mRNA represents up to 5% of total mRNA and AOX1 enzyme can be up to 30% of cell weight (Ref 5).

The EasySelect™ Pichia Expression system is supplied with the pPICZ and pPICZα vectors. The pPICZ and pPICZα vectors contain a multiple cloning site (MCS) and allow the insertion of your gene of interest under control of the AOX1 promoter. The pPICZ and pPICZα vectors also contain a c-Myc tag for easy detection and a polyhistidine (6xHis) tag for easy detection and purification of your protein.

Since Pichia pastoris naturally secretes few proteins, you can use the extracellular signaling sequence in the pPICZα vectors to force secretion of your protein into the culture medium to simplify down-stream purification.

Get High-Copy Number Integration with Zeocin™ Antibiotic
The EasySelect™ Pichia Expression Kit utilizes the Zeocin™ antibiotic to select for transformed Pichia cells. pPICZ and pPICZα vectors contain the bleomycin gene from Streptoalloteichus hindustanus (Sh ble) that confers resistance to Zeocin™ antibiotic. The pPICZ and pPICZα vectors can be stably integrated into the Pichia genome. By increasing the concentration of Zeocin™ antibiotic, you can easily select for clones with the highest number of integrants and highest protein expression levels.

The EasySelect™ Pichia Expression Kit contains the following components:
• Stab vials of Pichia pastoris and E. coli strains for starting your cultures.
• Solutions for making transformation-competent Pichia cells.
• pPICZ and pPICZα vectors for fusing your gene to the AOX1 promoter.
• Sequencing primers for confirming insertion of your gene in the pPICZ vectors.
• Zeocin™ antibiotic for selecting transformed Pichia cells.

For Research Use Only. Not intended for use in diagnostic procedures.

Related Links
View the product insert for the pPICZ A, B, C vectors (PDF).
View the product insert for the pPICZα A, B, C vectors (PDF).
View the product insert for Zeocin™ antibiotic (PDF).
Learn more about using Pichia pastoris in Pichia Protocols 1st edition available from Invitrogen.
Learn more about other Pichia expression systems from Invitrogen.

References

1. Cereghino JL, Cregg JM. Heterologous protein expression in the methylotrophic yeast Pichia pastoris. FEMS Microbiol Rev. 2000 Jan;24(1):45-66. [PubMed]
2. Cereghino GP, Cereghino JL, Ilgen C, Cregg JM. Production of recombinant proteins in fermenter cultures of the yeast Pichia pastoris. Curr Opin Biotechnol. 2002 Aug;13(4):329-32. [PubMed]
3. Cregg JM. Introduction: distinctions between Pichia pastoris and other expression systems. Methods Mol Biol. 2007;389:1-10. [PubMed]
4. Cregg JM, Tolstorukov I, Kusari A, Sunga J, Madden K, Chappell T. Expression in the yeast Pichia pastoris. Methods Enzymol. 2009;463:169-89. [PubMed]
5. Cregg JM, Madden KR, Barringer KJ, Thill GP, Stillman CA. Functional characterization of the two alcohol oxidase genes from the yeast Pichia pastoris. Mol Cell Biol. 1989 Mar;9(3):1316-23. [PubMed]
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Antibiotic Resistance (Bacterial): Zeocin™ (ZeoR)
Bacterial or Yeast Strain: GS115, KM71H, X-33
Cloning Method: Restriction Enzyme ⁄ MCS
Expression: Yeast
Expression Mechanism: Cell-Based Expression
Product Line: EasySelect™
Product Size: 1 kit
Promoter: AOX1
Protein Tag or Fusion: His Tag (6x), c-Myc Epitope Tag
Species: P. pastoris
Vector Type: pPIC

Contents & storage

The EasySelect Pichia Expression Kit includes 20 µg each of the plasmids pPICZ A, B, & C and pPICZα A, B, & C; Pichia pastoris strains X-33 (wild-type), GS115 (his4), KM71H (arg4 aox1::ARG4), and control strains for both intracellular and secreted expression; 250 mg Zeocin™ antibiotic; media (YP and YNB), EasyComp™ Pichia Transformation Kit, 2 µg each 5´ AOX1, AOX1, and α-factor sequencing primers. The pPICZ and pPICZα vectors are also available separately. A lacZ positive expression control is provided with pPICZ.

Store strains at +4°C. Store vectors and primers at -20°C. All components are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products