PureLink® Genomic DNA Mini Kit

Catalog number: K182000

Invitrogen™  Related applications: DNA Extraction | Epigenetic Sequencing | SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The PureLink® Genomic DNA Mini Kit enables high-yield, high-purity, genomic DNA (gDNA) extractions from a wide variety of sample types. Use this single kit for genomic DNA purification from blood, tissues, cells, bacteria, swabs, and blood spots, with a familiar silica-based, microcentrifuge spin-column format. Features of the PureLink® Genomic DNA Mini Kit include:

Kit flexibility—one kit works with a variety of sample types and sizes
Ultraclean gDNA—minimal contamination of DNA product means successful downstream application
Improved design—optimized spin-column design and buffer formulation for better yield and purity

Multiple sample sources, one kit
The PureLink® Genomic DNA Mini Kit can be used with many different sample types, each with its own optimized protocol outlined in the manual.

Optimized components and protocols
The PureLink® Genomic DNA Mini Kit includes parts and protocols that improve upon the easy and familiar spin-column methods currently in use (see figure). Columns are newly configured to bind and release DNA more efficiently (see figure). Each sample source uses a specialized lysis procedure and an optimized lysis buffer formulated to enhance proteinase K activity and eliminate protein contamination. A chaotropic salt buffer promotes stable binding of DNA to the column resin, and powerful wash buffers remove all traces of protein and salt. DNA is eluted in a low-salt buffer to allow for pH stabilization of the DNA in storage.

Higher yields and purity
With PureLink® Genomic DNA Kits you can expect high yields of high-purity gDNA (determined by A260/A280 measurements), no matter the sample type, from bacteria to tissues to blood to cultured cells (see figures). The enhanced PureLink® Lysis and Wash Buffers help to consistently deliver purified gDNA samples with absorbance reading ratios of ~1.9 for both A260/A280 and A260/A230, indicating no detectable contamination from carryover of protein or guanidine (see figure).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type (General): Bacteria, Cells, Tissue, Blood
Isolation Technology: Silica Spin Column
Downstream Application: PCR, Real-Time Quantitative PCR (qPCR), Sequencing, Southern Blotting, Genotyping
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Product Size: 10 preps
Shipping Condition: Room Temperature

Contents & storage

• 2.0 ml PureLink™ Genomic Lysis/Binding Buffer
• 1.8 ml PureLink™ Genomic Digestion Buffer
• 2.0 ml PureLink™ Genomic Wash Buffer 1
• 1.5 ml PureLink™ Genomic Wash Buffer 2
• 2.0 ml PureLink™ Genomic Elution Buffer
• 0.4 ml RNase A (20 mg/ml)
• 0.2 ml Proteinase K (20 mg/ml)
• 10 PureLink™ Spin Columns with Collection Tubes
• 20 PureLink™ Collecion Tubes (2.0 ml)

Store all components at room temperature.

Documents

Manuals & protocols

Recommended products