PureLink™ Genomic Plant DNA Purification Kit

Catalog number: K183001

Invitrogen™  Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The PureLink® Genomic Plant DNA Purification Kit allows efficient isolation of total DNA in about 40 minutes from a variety of plant samples, such as spinach, tomato, soy, wheat, Arabidopsis, and mushroom.
• Fast – Total DNA purification from difficult plant samples in under an hour
• Efficient – Yields high-quality DNA with minimal RNA contamination

Simple DNA Purification from Plant Samples
The PureLink® Genomic Plant DNA Purification Kit is suitable for isolating DNA from chloroplasts and a variety of plant tissues (Figure 1), including alfalfa sprouts, sunflower sprouts, corn husks, soybeans, mushroom, tomato leaves, wheat grass, and Arabidopsis thaliana leaves, in less than 40 minutes. The entire protocol relies on the centrifugation of spin columns in the absence of organic solvents. PureLink® Genomic Plant DNA Purification Kit is designed to purify DNA from up to 100 mg of plant tissue.

Maximize Yields of Total DNA
High yield of DNA and minimal DNA degradation is achieved by using young plant samples such as leaves and by freezing the sample in liquid nitrogen immediately after collection. Total DNA is eluted in 2 aliquots of 100 µl each to obtain greater DNA yield, with the first elution generating between 65-80% DNA recovery and the second elution > 90%. Typically, DNA isolated using the PureLink® Genomic Plant DNA Purification Kit has an A260/A280 > 1.80 when samples are diluted in Tris-HCl (pH 7.5), indicating that the DNA is reasonably free of proteins that could interfere with downstream applications.

Isolated DNA can be used for a Number of Downstream Applications:
Total DNA purification from different plant tissue with the PureLink® Genomic Plant DNA Purification Kit is suitable for a variety of applications, including PCR, restriction enzyme digestion, and Southern blotting.

For Research Use Only. Not intended for diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type (General): Fungi, Plant Samples
Isolation Technology: Silica Spin Column
Downstream Application: Cloning, PCR, Sequencing, Southern Blotting
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Product Line: PureLink™
Product Size: 50 preps

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products