S.N.A.P.™ Gel Purification Kit

Catalog number: K199925

Invitrogen™  Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The S.N.A.P.™ Gel Purification Kit is designed for rapid purification of DNA fragments from agarose gels. The key is the unique S.N.A.P.™ resin, which is provided ready to use in a spin-column format. The resin efficiently binds nucleic acids up to 20 kb and eliminates organic extractions, and the need to perform time-consuming ethanol precipitations.
Easy to Use
The S.N.A.P.™ Gel Purification Kit yields pure DNA in an easy-to-perform,
rapid procedure. Simply:
1. Excise DNA band from your agarose gel.
2. Add NaI solution (provided) and melt the agarose at 42-50°C.
3. Load sample onto the S.N.A.P.™ spin column.
4. Wash away contaminants.
5. Elute DNA in water.

The eluted DNA is highly pure and ready to use for cloning, sequencing, transformation, or transfection.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Downstream Application: Cloning, PCR
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
In-Gel Dye: Ethidium Bromide, SYBR® Safe
Isolation Technology: Spin Column
Product Line: S.N.A.P. ™
Product Size: 25 preps
Sample Type (General): Gel Samples
Sample Type (Specific): DNA Fragments in Agarose Gel Slices

Contents & storage

Each S.N.A.P.™ Gel Purification Kit includes enough S.N.A.P.™ columns, collection vials, and reagents for 25 gel purifications. Store all components at room temperature. All reagents are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols