TA Cloning® Kit, with pCR™2.1 Vector and One Shot® INVαF' Chemically Competent E. coli

Catalog number: K200001

Invitrogen™  Related applications: PCR Cloning

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The TA Cloning® Kit with pCR™2.1 vector provides a quick, one-step cloning strategy for directly inserting a Taq-amplified PCR product into a plasmid vector. The TA Cloning® Kit uses the pCR™2.1 cloning vector and ExpressLink™ T4 DNA Ligase to generate a ligation product in a fifteen-minute, room-temperature ligation step. Reactions typically yield >80% recombinants containing inserts.

Features of the TA Cloning® Kit with pCR™2.1 vector:
Fast & convenient—15-minute, room-temperature ligation
Efficient—blue/white screening and >80% clones with correct insert
Flexible—choice of kanamycin or ampicillin resistance for flexible antibiotic selection
Hassle-free—eliminates any enzymatic modifications of the PCR product
Streamlined—does not require the use of PCR primers that contain restriction sites

The pCR™2.1 vector provides:
• 3'-T overhangs for direct ligation of Taq-amplified PCR products
• T7 promoter for in vitro RNA transcription and sequencing
• A versatile polylinker with flanking EcoR I sites for easy excision of inserts
• M13 forward and reverse primer sites for sequencing

How TA Cloning® Works
Taq polymerase has a non-template-dependent activity that adds a single deoxyadenosine (A) to the 3' ends of PCR products. The linearized vector supplied in this kit has single 3' deoxythymidine (T) residues. This allows PCR inserts to ligate efficiently with the vector.

Kit Configurations
The TA Cloning® Kit is offered in a variety of configurations: without competent cells (K2020-20 and K2020-40), with One Shot® INVF' Chemically Competent E. coli (K2000-01 and K2000-40), with One Shot® TOP10F' Chemically Competent E. coli (K2030-01 and K2030-40), and with One Shot® TOP10 Chemically Competent E. coli (K2040-01 and K2040-40) in 20- and 40-reaction kit sizes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Vector Type: TA Cloning Vectors
Cloning Method: TA Cloning®
Bacterial or Yeast Strain: INVαF´
Product Size: 20 reactions

Contents & storage

TA Cloning® kits contain linearized pCR™2.1 vector, ExpressLink™ T4 DNA ligase, 5X ExpressLink™ T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls. The competent cell kits contain One Shot® chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.

Store One Shot® E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols