Invitrogen™

PureLink™ HiPure Plasmid Miniprep Kit

Catalog number:  K210003

 Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The PureLink® HiPure Plasmid Miniprep Kit allows isolation of high yields of highly pure plasmid DNA from E.coli in less than 2 hours, without the use of any organic solvents or cesium chloride, and contains low endotoxin levels.
• Fast – Highly-quality purified plasmid DNA in under 2 hours
• Efficient – Increased yields with low endotoxin levels

Plasmid DNA of High Purity
The PureLink® HiPure Plasmid Miniprep Kit is designed to efficiently isolate plasmid DNA from E.coli in 1.5-2 hours. The kit employs anion-exchange resin composed of small particles with a uniform pore size to purify plasmid DNA to a level equivalent to two passes through CsCl gradients (Figure 1). The need for additional steps to remove contaminants such as phenol, chloroform, and CsCl are eliminated, minimizing exposure to and disposal of hazardous materials.

High Yield and Efficient Endotoxin Removal
PureLink® HiPure Plasmid Miniprep Kit allows purification of up to 30 µg of high quality plasmid DNA from 1-3 ml overnight E.coli cultures in approximately 1 hour, with an endotoxin level of 0.1-1 EU/ µg, when cloning high copy number plasmids (Table 1). Typically, DNA isolated using the PureLink® HiPure Plasmid Miniprep Kit has an A260/A280 > 1.80 when samples are diluted in Tris-HCl (pH 7.5) indicating that the DNA is reasonably free of proteins that could interfere with downstream applications.

Isolated DNA can be used for a Number of Downstream Applications:
The purified DNA is ultrapure and suitable for downstream applications, including those requiring the highest purity DNA, such as mammalian transfections (Figure 2), automated and manual DNA sequencing, PCR amplification, in vitro transcription, bacterial cell transformation, cloning, and labeling.

For Research Use Only. Not intended for diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Sample Type (General): Bacteria
Downstream Application: Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing, Transfection, Transformation
Final Product: BAC DNA, Plasmid DNA
Isolation Technology: Silica Spin Column
Product Line: PureLink™
Product Size: 100 preps
Sample Type (Specific): Bacterial Culture
Shipping Condition: Room Temperature

Contents & storage

• 50 ml Resuspension Buffer (R3)
• 550 µl RNase A
• 50 ml Lysis Buffer (L7)
• 40 ml Precipitation Buffer (N3)
• 250 ml Equilibration Buffer (EQ1)
• 500 ml Wash Buffer (W8)
• 90 ml Elution Buffer (E4)
• 15 ml TE Buffer
• 100 HiPure Columns

Store all components at room temperature.

Documents

Manuals & protocols

Recommended products