PureLink™ HiPure Plasmid FP (Filter and Precipitator) Maxiprep Kit

Catalog number:  K210027

Invitrogen™  Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The PureLink™ HiPure Plasmid FP (Filter and Precipitator) Maxiprep Kit isolates plasmid DNA, from cell pellet to ultrapure DNA, in less than one hour. Using the HiPure Filter Columns and HiPure Precipitators supplied in the kit, you can purify plasmid DNA without the centrifugation steps required in traditional isolation protocols. The quality of the purified plasmid DNA is equivalent to that obtained by two passes through cesium chloride gradients - the most rigorous method for DNA purification - making it suitable for transfection and other applications requiring high-purity plasmid DNA. The PureLink™ HiPure Plasmid FP Maxiprep Kit (Table 1) provides:
• Simple protocols - bacterial lysate clarification and isopropanol precipitation without centrifugation
• Fast results - from E. coli cell pellet to high-purity, transfection-grade plasmid DNA in about 60 minutes
• High yields - up to 850 µg from a high-copy plasmid

How it works
The PureLink™ HiPure Plasmid FP Maxiprep Kit employs an alkaline lysis method to prepare and rapidly clear bacterial lysates without the need for centrifugation (Figure 1). The unique HiPure Filter Column includes a lysate filtration cartridge integrated into the anion-exchange column for single-step lysate clarification and plasmid DNA binding. Impurities such as RNA, proteins, and other contaminants are removed with a single wash, and ultrapure plasmid DNA is eluted with a high-salt buffer. To precipitate and desalt the DNA, isopropanol is added to the eluted DNA and the mixture is then applied to the HiPure Precipitator using a large syringe. After a subsequent washing and drying step, the plasmid DNA is easily eluted from the HiPure Precipitator with either TE buffer or water. The use of the precipitator eliminates the need for centrifugation, reducing the risk of losing the DNA pellet during supernatant removal, and saving time. The resulting DNA is ready to use in the most challenging applications. The entire protocol - from E. coli cell pellet to ultrapure transfection-grade plasmid DNA - can be completed in about an hour.

Highest yields and ready-to-use concentrations
The HiPure Precipitator has been optimized to allow for lower elution volumes (down to 500 µl) to produce highly concentrated DNA without a significant decrease in yields. In fact, yields at higher concentrations are consistently higher than the competitor kits (Figure 2). Yields and concentration can be adjusted to meet the needs of your downstream application.
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Sample Type (General): Bacteria
Downstream Application: Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing, Transfection, Transformation
Elution Volume - max: 15 ml
Elution Volume - min: 15 ml
Final Product: Plasmid DNA
Isolation Technology: Anion Exchange Resin with Lysate Clearing Filter & Precipitator
Number of Reactions: 25 Preps
Plasmid Type: <40kb, BAC, High Copy Plasmid, Low Copy Plasmid
Product Line: PureLink™
Product Size: 25 preps
Purification Time: 1 hr
Purity or Quality Grade: Transfection Grade
Quantity: 25 columns
Sample Type (Specific): Bacterial Culture
System: PureLink™
Yield (up to): 1000 µg

Contents & storage

The PureLink™ HiPure Plasmid FP Maxiprep Kit includes Equilibration Buffer, Resuspension Buffer, Lysis Buffer, Precipitation Buffer, RNase A, Wash Buffer, Elution Buffer, TE Buffer, HiPure Filter Columns, and a PureLink™ HiPure Precipitator Module. The PureLink™ HiPure Precipitator Module includes HiPure Precipitators, and 5 ml and 30 ml syringes. Store all components at room temperature.

Documents

Manuals & protocols

Recommended products