EveryPrep™ Universal Vacuum Manifold
Catalog number:  K211101
Invitrogen™  Related applications: DNA Extraction
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The EveryPrep™ Universal Vacuum Manifold consists of a dual chamber with interchangeable top plates that include a luer-lock tapped top plate, a 96-well top plate, a mini column elution plate and a maxi column elution plate.
The EveryPrep™ Universal Vacuum Manifold is designed for rapid manual purification of nucleic acids. It offers the ability to directly elute from the manifold using the new ChargeSwitch® Pro Filter Plasmid Mini, Midi and Maxi Kits and also many other competitors' vacuum assisted spin column preps and filter plates. The EveryPrep™ Universal Vacuum Manifold replaces and consolidates all previous manifolds into one space saving benchtop unit. The manifold reduces sample handling to a minimum by allowing direct parallel and simultaneous processing of up to 96 samples using collection plates, deep-well blocks, or microtubes. The use of a vacuum eliminates the need for time-consuming centrifugation. The manifold will consist of a dual chamber construct fashioned out of aluminum with interchangeable top plates of a luer-lock tapped top plate, a 96-well top plate, a mini column elution plate and a maxi column elution plate.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
LoyaltyEligible: SKUEligibleForLoyalty
Product Size: 1 each
Shipping Condition: Room Temperature
Type: Vacuum Manifold
Contents & storage
1 vacuum manifold. Store at room temperature.
Documents
Manuals & protocols