Zero Blunt® PCR Cloning Kit

Catalog number: K270020

Invitrogen™  Related applications: PCR Cloning

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Zero Blunt® PCR Cloning Kit offers an easy method for high-efficiency (>80%) cloning of blunt-end PCR products amplified with proof-reading, thermostable DNA polymerases. The Zero Blunt® PCR Cloning Kit uses the multipurpose cloning vector pCR™-Blunt and ExpressLink™ T4 DNA Ligase to generate a ligation product in a five-minute, room-temperature ligation step.

Features of the Zero Blunt® Cloning® Kit with pCR™-Blunt vector:
Fast & convenient—5-minute, room-temperature ligation
EfficientccdB gene for positive selection results in >80% clones with correct insert
Flexible—choice of kanamycin or Zeocin™ resistance for flexible antibiotic selection

The pCR™-Blunt vector provides:
EcoR I sites flanking the PCR product insertion site for excision of inserts
• T7 promoter/primer site for in vitro RNA transcription and sequencing
• M13 forward and reverse primer sites for sequencing or PCR screening

How Zero Blunt® PCR Cloning Works
The Zero Blunt® PCR Cloning Kit is designed to clone blunt PCR fragments (or any blunt DNA fragment) with a low background of non-recombinants. The pCR™-Blunt vector contains the lethal E. coliccdB gene fused to the C-terminus of LacZα (Bernard et al., 1994). Ligation of a blunt PCR fragment disrupts expression of the lacZα-ccdB gene fusion permitting growth of only positive recombinants upon transformation. Cells that contain non-recombinant vector are killed when the transformation mixture is plated.

Kit Configurations
The Zero Blunt® PCR Cloning Kit is offered in a variety of configurations: with One Shot® TOPO10 Chemically Competent E. coli (K2700-20 and K2700-40) and without competent cells (K2750-20 and K2750-40) in 20- and 40- reaction kit sizes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Vector Type: Blunt DNA Cloning Vectors
Cloning Method: Blunt PCR
Bacterial or Yeast Strain: TOP10
Product Size: 20 reactions

Contents & storage

Zero Blunt® PCR cloning kits contain linearized pCR™-Blunt vector, ExpressLink™ T4 DNA ligase , 5X ExpressLink™ T4 DNA ligase buffer, control template, dNTPs, sterile water, and M13 forward and reverse primers. Competent cell kits contain One Shot® chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.

Store the One Shot® E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products