Invitrogen™

Calcium Phosphate Transfection Kit

Catalog number: K278001
Invitrogen™

Calcium Phosphate Transfection Kit

Catalog number: K278001
Catalog Number
K278001
also known as K2780-01
Unit Size
75 rxns
Price (USD)
Price: 666.00
Online price: 631.65
Your price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K278001
also known as K2780-01
75 rxns
Price: 666.00
Online price: 631.65
Your price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Calcium Phosphate Transfection Kit provides high-quality reagents to enable the introduction of DNA into eukaryotic cells via calcium phosphate co-precipitation.

How it works
The calcium phosphate transfection method for introducing DNA into mammalian cells is based on forming a calcium phosphate-DNA precipitate. Calcium phosphate facilitates the binding of the DNA to the cell surface. DNA then enters the cell by endocytosis. The method was first developed by Graham and van der Ebb and was later modified by Wigler. The procedure is routinely used to transfect a wide variety of cell types for transient expression or for producing stable transformants. The DNA is mixed directly with a concentrated solution of CaCl2, which is then added dropwise to a phosphate buffer to form a fine precipitate. Aeration of the phosphate buffer while adding the DNA-CaCl2 solution helps to ensure that the precipitate that forms is as fine as possible, which is important because clumped DNA will not adhere to or enter the cell as efficiently.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Established Cell Lines
Format
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type
Plasmid DNA
Transfection Technique
Chemical Transfection
For Use With (Application)
Transfection
High-throughput Compatibility
Not High-throughput Compatible (Manual)
No. of Reactions
75 Reactions
Product Type
Transfection Kit
Quantity
75 Reactions
Serum Compatible
No

Contents & Storage

2 × 12 ml Tissue Culture Sterile Water
2 × 12 ml 2X HEPES Buffered Saline (HBS)
3 × 1 ml 2 M CaCl2
20 μg pcDNA™3.1/His/lacZ

Store all components at -5 to -30°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text