Zero Blunt® TOPO® PCR Cloning Kit, with One Shot® TOP10 Chemically Competent E. coli cells

Catalog number: K280020

Invitrogen™  Related applications: PCR Cloning

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Zero Blunt® TOPO® PCR Cloning Kit provides a highly efficient, 5-minute, one-step cloning strategy ("TOPO® Cloning") for the direct insertion of blunt-ended PCR products amplified with proofreading thermostable polymerases into a plasmid vector. Each kit has the Zero Blunt® TOPO® vector (see figure) containing the ccdB gene for positive selection, only permitting growth of plasmid vectors with recombinants. Zero® Blunt® TOPO® kits are available with a variety of competent cells, or without competent cells, depending on your needs and budget. Features of the Zero® Blunt® TOPO® PCR Cloning Kits:

Fast and easy—go from PCR to clones in just 3 steps and in as little as 5 minutes hands-on time
Efficient—obtain up to 95% clones with correct insert
Proven—reliable performance for over a decade with over 4,000 citations
Simple —no ligase, post-PCR procedures, or PCR primers containing specific sequences are required.

Zero Blunt® TOPO® Cloning Kits—overview

Vector: pCR™Blunt II-TOPO® vector—contains the ccdB gene, allowing for direct selection of recombinants

Cloning method: TOPO® cloning—Topoisomerase I–based, 5-minute ligation of blunt-ended PCR products amplified with proofreading thermostable polymerases

Competent cells: Various options—choose from kits with either general, high-efficiency, bacteriophage T1-resistant, fast-growing competent cells, or use your own.

pCR™Blunt II-TOPO® vector—designed for proofreading polymerases
The pCR™Blunt II-TOPO® vector is designed to clone blunt-ended PCR products generated by thermostable proofreading polymerases such as Platinum® Pfx DNA Polymerase. The pCR™Blunt II-TOPO® vector contains:

• EcoRI sites flanking the PCR product insertion site for easy excision of inserts
• Kanamycin and Zeocin™ resistance genes for your choice of selection in E. coli
• SP6 promoter/primer site for in vitro RNA transcription and sequencing
• M13 forward and reverse primer sites for sequencing or PCR screening

pCR™Blunt II-TOPO® clone selection
pCR™-Blunt II-TOPO® allows for direct selection of recombinants via disruption of the lethal E. coli gene, ccdB. The vector contains the ccdB gene fused to the C-terminus of the LacZα fragment. Ligation of a blunt-end PCR product disrupts expression of the LacZα-ccdB gene fusion, permitting growth of only positive recombinants upon transformation. Cells that contain non-recombinant vector are killed upon plating. Therefore, blue/white screening is not required.

Simplified TOPO®-based cloning
Using TOPO® cloning technology, there is no need for PCR primers containing specific sequences, post-PCR procedures, vector preparation, or other time-intensive DNA manipulation steps. Just add your PCR reaction straight to the provided topoisomerase-charged vector, incubate 5 minutes, and transform E. coli competent cells.

Efficient cloning
With up to 95% of clones carrying the desired insert, you can screen less clones, saving you time and money. The pCR™Blunt II-TOPO® vector used in this kit comes with lethal ccdB gene for efficient selection of blunt-ended PCR recombinants products generated bythermostable proofreading polymerases.

The standard in cloning
When it comes to cloning, TOPO® cloning technology has been a reliable partner for thousands of scientists for over ten years. Fast, simple-to-use, and efficient, TOPO® cloning has been applied to many different vectors for a wide array of applications.

Zero Blunt® TOPO® PCR Cloning® Kit options
The Zero Blunt® TOPO® PCR Cloning Kit for direct insertion of blunt-ended PCR products into a plasmid vector can be purchased with a variety of competent cells that deliver different advantages depending upon your needs:

General cloning: TOP10 cells (Cat. No. K2800-20, K2800-40)
High-efficiency cloning: TOP10 Electrocomp™ Cells (Cat. No. K2860-01, K2860-40)
General cloning, bacteriophage T1 resistance: DH5α-T1R (Cat. No. K2820-20, K2820-40)
Fast growth: Mach1™-T1R Chemically Competent E. Coli (Cat. No. K2830-20)
Provide your own: for flexibility and to save money (Cat. No. 450254)

We also offer a version of the kit that includes a PureLink® Quick Plasmid Miniprep Kit (Cat. No. K2800-02) for use in isolation of clean, sequencing-ready, recombinant plasmid.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Vector Type: Blunt DNA Cloning Vectors
Cloning Method: Blunt TOPO®
Bacterial or Yeast Strain: TOP10
Product Size: 25 reactions

Contents & storage

Box 1:
• Linearized and topoisomerase I-activated pCR™Blunt II-TOPO® vector
• Salt solution
• dNTPs
• Control template
• M13 forward and reverse primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.

Box 2:
• One Shot® Chemically Competent or Electrocomp™ E. coli
• S.O.C. medium
• Supercoiled pUC19 control plasmid

Store at -68 to -85°C.

Documents

Manuals & protocols

Recommended products