PureLink™ PCR Purification Kit

Catalog number: K310002

Invitrogen™  Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The PureLink PCR Purification Kit provides a more convenient, rapid, and high-yielding purification process compared to other companies' kits. After PCR amplification, purification of the PCR amplicons is required for many common downstream applications, including enzyme digestion, ligation, sequencing, and labeling experiments, to prevent enzymes (polymerase, restriction enzyme) and reaction components (dNTPs, primers, buffers, salts) from carrying over, contaminating, and negatively influencing the downstream experiments.

Advantages of the PureLink PCR Purification Kit include:
• 4X binding capacity compared to Company Q; 40 µg for the PureLink kit versus 10 µg from other companies' kits
• No need for tedious sample pH adjustments necessary with other kits
• Convenient and rapid protocol, completed in less than 10 minutes
• Optional size-selection buffers included for selectively purifying 300– 600 bp fragments or removing fragments < 300 bp

The PureLink PCR Purification Kit is based on the selective binding of dsDNA to silica-based membrane in the presence of chaotropic salts. Simply mix the PCR product with Binding Buffer and apply to the PureLink Spin Column. The dsDNA binds to the silica-based membrane in the column. Remove impurities by thorough washing with Wash Buffer. To purify the DNA, elute the dsDNA in low-salt Elution Buffer or water. The purified PCR product is suitable for automated fluorescent DNA sequencing, restriction enzyme digestion, and cloning.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

High Throughput Compatibility: High Throughput-Compatible
Sample Type (General): Enzymatic Reactions (e.g. PCR)
Downstream Application: Cloning, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing
Isolation Technology: Silica Spin Column
Product Line: PureLink™
Product Size: 250 preps
Sample Type (Specific): PCR Products
Shipping Condition: Room Temperature

Contents & storage

• 72 ml Binding Buffer (B2)
• 109 ml Binding Buffer High-Cutoff (B3)
• 80 ml Wash Buffer (W1)
• 15 ml Elution Buffer (E1)
• 5 × 50 PureLink® PCR Spin Columns with Collection Tubes
• 5 × 50 PureLink® Elution Tubes (1.7 ml)

Store at room temperature.

Documents

Manuals & protocols

Recommended products