PureLink™ PCR Purification Binding Buffer B2

Catalog number: K310005

Invitrogen™  Related applications: Automated Nucleic Acid Purification | DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

PureLink™ PCR Purification Binding Buffer B2 is the binding buffer used with the PureLink™ PCR Purification Kit. It is added directly to PCR products in the first step of the PureLink™ PCR Purification Kit procedure to facilitate DNA binding to the silica membrane. The buffer is activated by the addition of 80mls of Isopropanol.

The PureLink™ PCR Purification Kit is used for purifying PCR products ranging in size from 100 bp-12kb.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer Type: Binding Buffer
Product Size: 120 mL
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols