pBAD/TOPO™ ThioFusion™ Expression Kit

Catalog number: K37001

Invitrogen™  Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The pBAD/TOPO® ThioFusion™ Expression Kit is designed for one-step cloning and regulated expression of thioredoxin fusion proteins in E. coli(Figure 1). The pBAD/Thio-TOPO® vector encodes His-Patch thioredoxin as an N-terminal fusion partner. The thioredoxin fusion can significantly increase the solubility of many difficult-to-express proteins and improve the yield of protein production. The vector includes the following additional features (Figure 2):

• The araBAD promoter for tightly regulated expression in E. coli I-activated vector for 5-minute TOPO® Cloning of Taq-amplified PCR products
• C-terminal polyhistidine (6xHis) tag for purification with nickel-chelating resin and detection with an Anti-His(C-term) Antibody
• C-terminal V5 epitope for detection with an Anti-V5 Antibody
• Enterokinase cleavage site for efficient cleavage of N-terminal fusion tag with EKMax™ Enterokinase
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

Expression: E. coli
Promoter: araBAD
Bacterial or Yeast Strain: TOP10
Antibiotic Resistance (Bacterial): Ampicillin (AmpR)
Cleavage: EK (Enterokinase) Recognition Site
Cloning Method: TOPO®-TA
Constitutive or Inducible System: Inducible
Expression Mechanism: Cell-Based Expression
Inducing Agent: Arabinose
Product Line: TOPO®, ThioFusion™
Product Size: 20 reactions
Protein Tag or Fusion: His Tag (6x), Thioredoxin, V5 Epitope Tag
Selection Agent (Eukaryotic): None
Vector Type: pBAD

Contents & storage

Each pBAD/TOPO® ThioFusion™ Expression Kit contains two boxes and the LMG194 E. coli stab. The pBAD/TOPO® ThioFusion™ Expression box contains all of the reagents necessary for PCR (except Taq polymerase) including 200 ng of topoisomerase I-activated pBAD/Thio-TOPO® vector, sterile water, dNTPs, 10X PCR buffer, salt solution, control template and primers, 20% L-arabinose, primers for sequencing or PCR screening, and an expression control plasmid. Store at -20°C. The One Shot® box contains all of the reagents necessary for transformation including 21 single-use 50 µl aliquots of chemically competent TOP10 E. coli, S.O.C. medium, and pUC19 supercoiled control plasmid. Store the One Shot® box at -80°C. Store the LMG194 E. coli stab at 2 - 8°C. All components are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products