Related applications: Gateway Cloning | RNAi
Error loading your content!
Catalog number |
Select a plan
Unit size
|
Price ({{currency}}) | Availability | Qty | |
---|---|---|---|---|---|
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} |
|
— |
Cloning Method: | Gateway® |
---|---|
Constitutive or Inducible System: | Inducible |
Delivery Method: | Transfection |
Product Line: | BLOCK-iT™, Gateway® |
Product Size: | 20 reactions |
Promoter: | H1⁄TO |
RNAi Type: | shRNA |
Selection Agent (Eukaryotic): | Zeocin™ |
Vector Type: | BLOCK-iT RNAi Vectors |