BLOCK-iT™ U6 RNAi Entry Vector Kit

Numéro de catalogue:  K494500

Invitrogen™  Applications liées: Gateway Cloning | RNAi

Error lors du chargement de votre contenu!

  Numéro de catalogue
Choisir votre mod�le de prix
Taille unitaire
Prix ({{currency}}) Disponibilité Qté
{{product.sku}} {{product.sku}} également connu comme {{product.formattedSku}} 
{{product.availability.message}}
Ajouts

Votre stock sur site

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} En stock)
›› {{supplyCenter.scName}}(En rupture de stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Haut de la page

Description

The BLOCK-iT™ U6 RNAi Entry Vector provides a simple, streamlined approach to cloning short hairpin RNA (shRNA) sequences for testing in transient transfections for RNA interference (RNAi), a preferred technique for analyzing gene downregulation. An easy cloning process places an ~50-bp oligonucleotide DNA immediately following a U6 pol III type promoter (Figure 1). Expression of this RNAi cassette forms a shRNA molecule in the cell that will be processed and act as short interfering RNA (siRNA) that will generate the RNAi effect.

Delivery of the U6 RNAi Cassette
Once cloning is complete, the U6 Entry Vector is ready to be used immediately in transient transfections using a reagent such as Lipofectamine™ 2000. This makes this system ideal for initial shRNA screenings in many mammalian cell types. As an alternative in hard-to-transfect or non-dividing cell types, or to deliver to animal model systems, the RNAi cassette can easily be recombined into a BLOCK-iT™ Viral RNAi vector. For stable shRNA lentiviral delivery and expression, recombine the BLOCK-iT™ U6 Entry Vector with the pLenti6/BLOCK-iT™ RNAi Vector. For transient delivery to challenging cell types using adenoviral transduction, recombine with the pAd/BLOCK-iT™ RNAi Vector.
For Research Use Only. Not for use in diagnostic procedures.

Spécifications

Cloning Method: Gateway®
Constitutive or Inducible System: Constitutive
Delivery Method: Transfection
Product Line: BLOCK-iT™
Product Size: 20 constructions
Promoter: U6
RNAi Type: shRNA
Vector Type: BLOCK-iT RNAi Vectors

Contenu et stockage

The BLOCK-iT™ U6 Entry Vector Kit contains two boxes. The cloning box contains four tubes of linearized vector at 5 ng/tube, enough to complete 5 cloning reactions per tube. Also included are 250 µl 10X annealing buffer, 20 µl T4 DNA ligase, 80 µl 5X DNA ligase buffer, 1.4 ng lacZ control oligo, 10 µl lyophilized lacZ control plasmid, 1.5 ml DNase/RNase-free water, and 2 µg each of forward and reverse sequencing primer. Store the vectors, buffers, control oligo and plasmid, water, and sequencing primers at -20°C. The One Shot® box contains transformation reagents including twenty-one 50-µl aliquots of One Shot® TOP10 Chemically Competent E. coli, S.O.C. medium, and a pUC19 supercoiled control plasmid. Store at -80°C. All reagents are guaranteed stable for 6 months when properly stored.

Documents

Manuels et protocoles

Produits recommandés