Invitrogen™
S.c. EasyComp™ Transformation Kit
Catalog number:  K505001
 Related applications: Protein Expression | Transformation
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The S.c. EasyComp™ Transformation Kit is designed for rapid preparation of transformation-competent Saccharomyces cerevisiae cells. Advantages over commonly used transformation procedures such as spheroplast formation or LiCl methods include:
• Cells can be made competent in less than 30 minutes
• Competent cell preparation and transformation solutions are optimized for use with S. cerevisiae
are provided ready-to-use and tested to ensure successful yeast transformation
• Competent cells can be frozen for future use
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Line: EasyComp™
Product Size: 1 kit
Target Organism Class: S. cerevisiae
Contents & storage
The S.c. EasyComp™ Transformation Kit provides enough reagents to prepare 6 batches of competent cells. Each batch is sufficient for 20 transformations. The kit should be stored at +4°C. All components are guaranteed stable for 6 months when properly stored.
Documents
Manuals & protocols
Recommended products