pSecTag/FRT/V5-His© TOPO® TA Expression Kit

Catalog number: K602501

Invitrogen™  Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Using restriction enzymes to clone your gene into an expression vector often forces you to compromise the final sequence of your insert (Figure 1A), especially when there are no useful restriction sites close to your genes coding sequence. This may result in suboptimal spacing of expression elements or incorporation of non-native amino acid residues, which can reduce your expression levels and/or cause the production of non-functional protein.

In addition to being a more effective way to clone, TOPO® Cloning eliminates these potential expression problems. TOPO® Expression Vectors enable you to insert the exact DNA sequence you require simply by performing PCR with appropriately designed primers. Your PCR product is cloned at a high efficiency in only five minutes into a topoisomerase I-activated expression vector. The resulting recombinant expression vector contains your exact DNA sequence without any non-coding regions (Figure 1B).

Many of Invitrogen's powerful expression vectors are available adapted for one-step TOPO® cloning and expression of PCR products. In addition, several expression vectors are now adapted for Directional TOPO® Cloning, enabling you to use proofreading polymerases and to clone your PCR products in a specific orientation.
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

Promoter: CMV
Key Function: Secreted Expression
Protein Tag or Fusion: His Tag (6x), IgK Leader Sequence, V5 Epitope Tag
Delivery Method: Transfection
Cloning Method: TOPO®-TA
Selection Agent (Eukaryotic): Hygromycin
Vector Type: TOPO-TA Vectors
Constitutive or Inducible System: Constitutive
Product Line: Flp-In™, TOPO®
Product Size: 20 reactions

Documents

Manuals & protocols