Champion™ pET300/NT-DEST and pET301/CT-DEST Gateway™ Vector Kit

Numéro de catalogue:  K630001

Invitrogen™  Applications liées: Protein Expression

Error lors du chargement de votre contenu!

  Numéro de catalogue
Choisir votre mod�le de prix
Taille unitaire
Prix ({{currency}}) Disponibilité Qté
{{product.sku}} {{product.sku}} également connu comme {{product.formattedSku}} 
{{product.availability.message}}
Ajouts

Votre stock sur site

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} En stock)
›› {{supplyCenter.scName}}(En rupture de stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Haut de la page

Description

The Champion™ pET300/NT-DEST and pET301/CT-DEST Gateway® Vector Kit is designed for rapid cloning with a Gateway® entry clone and subsequent high-level prokaryotic expression controlled by the strong bacteriophage T7 promoter. In addition to the T7 promoter, each vector contains only the necessary functional elements and an N- or C-terminal 6xHis tag (pET300/NT-DEST and pET301/CT-DEST, respectively) for convenient purification and detection (Figure 1). The vector kit is ideal for structural biologists who desire no or minimal modifications to their protein of interest. To maximize expression, use with MagicMedia™ E. coli Expression Medium.



Contents and Storage:
The Champion™ pET300/NT-DEST and pET301/CT-DEST Gateway® Vector Kit includes 6 µg each of pET300/NT-DEST and pET301/CT-DEST vectors and 10 µg of control vector. Store at -20“C. Guaranteed stable for 6 months when properly stored.
For Research Use Only. Not for use in diagnostic procedures.

Spécifications

Vector Type: pDEST, pET
Cloning Method: Gateway®
Promoter: T7
Protein Tag or Fusion: His Tag (6x)
Antibiotic Resistance (Bacterial): Ampicillin (AmpR)
Constitutive or Inducible System: Inducible
Inducing Agent: IPTG
Product Line: Champion™, Gateway®
Product Size: 6 µg
Selection Agent (Eukaryotic): None

Documents

Manuels et protocoles