FreeStyle™ MAX 293 Expression System

Catalog number:  K900010

Invitrogen™  Related applications: Antibody & Cell Culture Production | Protein Expression | Transfection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The FreeStyle™ MAX System is a breakthrough technology for rapid and high-yield mammalian protein production. This new mammalian expression technology is available in two convenient formats. FreeStyle™ MAX Reagent is a novel cationic lipid-based reagent designed to transfect FreeStyle™ cells with high efficiency. The FreeStyle™ MAX Expression System combines GIBCO® FreeStyle™ Media, FreeStyle™ MAX Reagent, and either FreeStyle™ CHO-S cells or FreeStyle™ 293 cells. The only equipment required to use the FreeStyle™ MAX System is a CO2 incubator and a platform shaker with shake flasks or a spinner culture.

The FreeStyle™ MAX System provides:
• An easy, rapid protocol for transient transfection of CHO and 293 cells in suspension cultures
• Functional proteins (up to milligram quantities) with all post-translational modifications produced in one week
• Simplified downstream purification of secreted proteins with FreeStyle™ serum-free medium
• Straightforward protocol for production scale-up

The FreeStyle™ MAX System can replace time-consuming stable cell line generation with large-scale functional protein production in one week.

Contents and Storage:
Store FreeStyle™ cells in liquid nitrogen. Store FreeStyle™ MAX Reagent at +4°C. Store GIBCO® FreeStyle™ CHO Expression Medium, FreeStyle™ 293 Expression Medium, and OptiPRO™ SFM at +2-8°C in the dark. Store the pCMV SPORT at -20°C. Stability is guaranteed for 6 months under the recommended storage conditions. Please refer to the manual for additonal storage information.
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

LoyaltyEligible: SKUEligibleForLoyalty
Cell Line: FreeStyle™ 293-F Cells
Classification: Animal Origin-Free, Serum-Free
Culture Type: Suspension Cell Culture
Delivery Method: Transfection
Expression: Mammalian
Expression Mechanism: Cell-Based Expression
Key Function: Protein Production
Product Line: Freestyle™
Product Size: 1 kit

Documents

Manuals & protocols

Recommended products