Ni-NTA Purification System
Catalog number: K95001
Novex™  Related applications: Protein Purification & Isolation
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The Ni-NTA Purification System is designed for the purification of recombinant proteins that contain a polyhistidine (6xHis) sequence. The kit utilizes Ni-NTA nickel-chelating resin and is supplied with native and denaturing buffers for efficient purification of recombinant proteins under different conditions.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format: Suspension
Column Type: Affinity Column
Product Size: 6 purifications
Immobilized Phase: Ni-NTA
Contents & storage
Six 2-ml resin columns and buffers for native and denaturing purification. Ten milliliters of Ni-NTA Agarose. Store at +4°C. All reagents are guaranteed stable for 6 months when properly stored.
Documents
Manuals & protocols
Recommended products