LIF Human ELISA Kit

Catalog number: KAC1351

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The LIF Human ELISA Kit is designed to quantify human LIF protein levels in serum, plasma, supernatant, and other biological fluids. Leukemia inhibitory factor (LIF) has pleiotropic functions and is produced by lymphocytes and macrophages.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gene Symbol: LIF
Assay Incubation Time: 4.5 hr
Assay Range: 300-9000 pg/mL
Assay Sensitivity: 20 pg/mL
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Gene Aliases: CDF, DIA, HILDA, MLPLI
Gene ID: 3976
Gene Name: leukemia inhibitory factor
Immunoassay Kit Format: Coated ELISA Kit
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 96 tests
Protein Aliases: LIF, Differentiation-stimulating factor, D factor, Melanoma-derived LPL inhibitor, MLPLI
Protein Name: Leukemia inhibitory factor
Sample Type (Specific): Cell Culture Supernatants, Plasma, Serum
Sample Volume: 100 µL
Shipping Condition: Wet Ice
Target (Kit Named): LIF
Target Species (Named): Human
Target Species (Validated): Human
UniProt ID: P15018

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody(s), diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols