MMP3 Human ELISA Kit

Catalog number: KAC1541

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The MMP-3 Human ELISA Kit is designed to quantify protein levels of human MMP-3 in serum, plasma, supernatant, and synovial fluid. Matrix metalloproteinase-3 (MMP-3) degrades connective tissue matrix.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Incubation Time: 3 hr
Assay Range: 0.62-20 ng/mL
Assay Sensitivity: <0.1 ng/mL
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Former Identifiers: Novex
Gene Name: matrix metallopeptidase 3
Immunoassay Kit Format: Coated ELISA Kit
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 96 tests
Protein Aliases: SL-1, Matrix metalloproteinase-3, MMP-3, Transin-1
Protein Name: Stromelysin-1
Protein Subtype: Proteases
Sample Type (Specific): Cell Culture Supernatants, Plasma, Serum, Synovial Fluid
Sample Volume: 50 µL
Shipping Condition: Wet Ice
Target (Kit Named): MMP3
Target Species (Named): Human
Target Species (Validated): Human
UniProt ID: P08254

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols