G-CSF Human ELISA Kit

Catalog number: KHC2031

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The G-CSF Human ELISA Kit quantifies natural and recombinant human Granulocyte Colony Stimulating Factor (G-CSF) in serum, plasma, buffered solution, or cell culture medium. G-CSF supports growth of hematopoietic colonies.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Cytokines & Receptors, Growth Factors & Receptors
Assay Incubation Time: 3 hr
Assay Range: 39-2500 pg/mL
Assay Sensitivity: <20 pg/mL
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Former Identifiers: Novex
Gene Aliases: C17orf33OS, GCSF, CSF3
Gene ID: 1440
Gene Name: colony stimulating factor 3
Gene Symbol: CSF3
Immunoassay Kit Format: Coated ELISA Kit
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 96 tests
Protein Aliases: G-CSF, Pluripoietin
Protein Name: Granulocyte colony-stimulating factor
Sample Type (Specific): Cell Culture Supernatants, Plasma, Serum
Sample Volume: 50 µL
Shipping Condition: Wet Ice
Target (Kit Named): G-CSF
Target Species (Named): Human
Target Species (Validated): Human
UniProt ID: P09919

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols