IGF1R (Phospho) [pY1135/pY1136] Human ELISA Kit

Catalog number: KHO0501

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The IGF-1R [pYpY1135/1136] Human ELISA kit is designed to detect and quantify the level of IGF-1R protein phosphorylated at tyrosine residues 1135 and 1136. This assay is intended for the detection of IGF-1R [pYpY1135/1136] from human, rat and mouse cell extracts using 96-well plates and a microplate reader.

Easy to run
More sensitive than Westerns
Convenient precoated, removable 8-well strips

Easy-to-Run Sandwich ELISA
A monoclonal capture antibody specific for IGF-1R has been coated onto the wells of the 96-well plate provided. During the first incubation, standards of known IGF-1R [pYpY1135/1136] content and unknown samples are pipetted into the wells and the IGF-1R antigen binds to the immobilized (capture) antibody. After washing, a rabbit antibody specific for IGF-1R phosphorylated at tyrosine 1135 1136 is added to the wells. During the second incubation, this antibody serves as a detection antibody by binding to the immobilized IGF-1R protein captured during the first incubation. After washing, a horseradish peroxidase labeled anti-rabbit IgG is added. This binds to the detection antibody to complete the four member sandwich. After a third incubation and washing to remove all the unbound enzyme, a substrate solution (TMB) is added, which is acted upon by the bound enzyme to produce color. The intensity of this colored product is directly proportional to the concentration of IGF-1R [pYpY1135/1136] present in the original specimen and the optical density can be read on a standard microplate reader. The total assay incubation time is only 4 hours.

Detect Low Expressing Proteins and Use Less Sample
ELISA kits designed to measure intracellular signaling targets are 2–10X more sensitive than Western blotting. The improved sensitivity enables you to detect low-expressing proteins that otherwise may not be distinguishable from background. In addition, the amount of sample needed to run the assay is less than what is needed for Western blots.

Ready-to-Use IGF-1R [pYpY1135/1136] Human ELISA Kit Contains These Reagents, Buffers, and Supplies:

• IGF-1R Antibody-Coated 96-Well Plate
• IGF-1R [pYpY1135/1136] Standard
• Standard Diluent Buffer
• IGF-1R [pYpY1135/1136] Detection Antibody
• anti-rabbit IgG-HRP (100X)
• HRP Diluent
• Wash Buffer Concentrate (25X)
• Stabilized Chromogen, TMB
• Stop Solution
• Plate Covers
• Detailed protocol with validation tests

Convenient Kit Format
The kit comes with a capture antibody precoated to the plate, eliminating an overnight coating process. The plates are machine coated to provide low variability well-to-well. Furthermore, the plate comes as removable 8-well strips allowing you to run as few samples as you want. In addition, the standards provided in the kit can be used to obtain quantitative results from the standard curve and/or can be used as a positive control.

Rigorous Validation
Each ELISA kit is validated for sensitivity, specificity, precision, and lot-to-lot consistency. See protocol insert for more information on validation.

Part of the ELISA Kit Family
We have a wide array of ELISA kits targeted to disease-related proteins. With excellent sensitivity, proven specificity and consistent performance, these optimized ELISA kits offer you a choice when it comes to getting physiologically relevant and quantitative results.

Learn more about ELISA kits.
Learn more about other immunoassays.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Incubation Time: 4 hr
Assay Range: 1.6 U/mL
Assay Sensitivity: <1 U/mL
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Former Identifiers: Novex
Gene Aliases: CD221, IGFIR, IGFR, JTK13
Gene ID: 3480
Gene Name: insulin like growth factor 1 receptor
Gene Symbol: IGF1R
Immunoassay Kit Format: Coated ELISA Kit
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 96 tests
Protein Aliases: Insulin-like growth factor I receptor, IGF-I receptor
Protein Name: Insulin-like growth factor 1 receptor
Protein Subtype: Receptor Tyrosine Kinases
Sample Type (Specific): Cell Extracts
Sample Volume: 10 µL
Shipping Condition: Wet Ice
Target (Kit Named): IGF1R
Target Site-Specificity: [pY1135/pY1136]
Target Species (Named): Human
Target Species (Validated): Human, Mouse, Rat
Target Sub-Specificity: Phospho
UniProt ID: P08069

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols