STAT5 alpha (Phospho) [pY694] Human ELISA Kit

Catalog number: KHO0761

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The STAT5a [pY694] Human ELISA Kit is designed to detect and quantify the levels of STAT5a phosphorylated at site tyrosine 694. Signal Transducer and Activator of Transcription 5 (STAT5) regulates gene transcription.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Incubation Time: 4 hr
Assay Range: 1.6 U/mL
Assay Sensitivity: <1 U/mL
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Former Identifiers: Novex
Gene Aliases: MGF, STAT5
Gene ID: 6776
Gene Name: signal transducer and activator of transcription 5A
Gene Symbol: STAT5A
Immunoassay Kit Format: Coated ELISA Kit
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 96 tests
Protein Name: Signal transducer and activator of transcription 5A
Sample Type (Specific): Cell Extracts
Sample Volume: 10 µL
Shipping Condition: Wet Ice
Target (Kit Named): STAT5 alpha
Target Site-Specificity: [pY694]
Target Species (Named): Human
Target Species (Validated): Human
Target Sub-Specificity: Phospho
UniProt ID: P42229

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols