Arcturus® Paradise® PLUS 1.5 Round Kit

Catalog number: KIT0301

Applied Biosystems™  Related applications: Laser Capture Microdissection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The ARCTURUS® Paradise® PLUS Reagent System permits unprecedented gene expression analysis of millions of previously inaccessible samples. It is an integrated solution for gene expression studies from formalin-fixed, paraffin-embedded (FFPE) tissues, including reagents for tissue staining, RNA isolation and RNA amplification. Amplify RNA from as little as 5 ng of fixed RNA using methods that have been specially optimized to overcome the challenges of cross-linked templates.

Key product features:
• Accelerated discovery process – unlock the tissue archives to access previously inaccessible samples
• Simplified procedure – achieve unprecedented results in six steps
• Streamlined application – integrate seamlessly into current workflow
• Efficient workflow – secure downstream success with the Paradise® QC Kit
• Comprehensive system – benefit from the only complete system for formalin-fixed samples

Unlock the Tissue Archives
Currently, tens of millions of tissue samples with known outcomes are available in tissue archives. The Paradise® PLUS Reagent System enables microarray and qRT-PCR analysis of these previously inaccessible samples, allowing retrospective studies to accelerate discoveries.

Achieve Unprecedented Results in Six Steps
Only six steps are required to take you from sample to results (See Figure 1).

Integrate Seamlessly into Current Workflow
The Paradise® PLUS Reagent System is compatible with all major microarray platforms. Robust and reproducible results are achieved using tools and techniques commonly employed in today’s research laboratories (See Figures 2, 3, 4, and 5).

Secure Downstream Success with the Paradise® QC Kit
Not all FFPE samples contain high quality RNA. Identify the best samples for your studies using a simple, real-time PCR research technique, and then proceed with confidence using these pre-qualified samples (See Figure 6).

Benefit from the Only Complete System for Formalin-fixed Samples
The Paradise® PLUS Reagent System is the only available solution that provides all the components necessary to prepare RNA from formalin-fixed, paraffin-embedded tissue samples for gene expression analysis. To ensure successful microarray results, the system includes reagents optimized for exceptional recovery of RNA and superior RNA amplification.

Integrated Systems for Microgenomics
The Paradise PLUS Reagent System is a validated as part of the ARCTURUS® Complete System for Microgenomics®, an integrated solution for utilizing small quantities of RNA for gene expression analysis. This system features the ARCTURUSXT™ Laser Capture Microdissection (LCM) Instrument to procure pure cell populations. ARCTURUS® Systems for Microgenomics enable accurate and sensitive microarray assays that reveal molecular signatures otherwise obscured in whole tissue samples or cell mixtures.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Downstream Application: Microarray Analysis, Real-Time Quantitative PCR (qPCR)
For Use With (Equipment): ArcturusXT™ LCM Instrument
Product Line: ARCTURUS®, Applied Biosystems™, Paradise®
Product Size: 48 samples
Sample Type (General): FFPE & Fixed Samples
Sample Type (Specific): Tissue (FFPE)
Workflow Step: In Vitro Transcription, RNA Amplification, RNA Extraction, Reverse Transcription, Tissue Staining

Contents & storage

Paradise Staining Kit (stain, staining jars, dehydration reagents), RNA Isolation Kit (RNA extraction and purification reagents, purification columns), RNA Amplification (cDNA synthesis and IVT modules), Amplification Purification Kit (purification reagents and columns). Variable storage conditions.

Documents

Manuals & protocols