Arcturus™ Paradise™ PLUS QC Kit

Catalog number:  KIT0303

Applied Biosystems™  Related applications: Laser Capture Microdissection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Formalin-fixed paraffin-embedded (FFPE) samples introduce unique challenges for gene expression profiling and gene expression quantitation, including chemical modification and fragmentation of RNA molecules. Archival FFPE samples present additional challenges due to increased RNA degradation over time. As a result, not all FFPE samples contain high quality RNA.

The ARCTURUS® Paradise® PLUS Sample Quality Assessment Kit offers a simple solution for evaluating the quality of RNA from Formalin-Fixed Paraffin-Embedded (FFPE) tissues before proceeding with laser microdissection and downstream molecular analysis. The reagents provided in the kit are optimized to enable efficient extraction, isolation, and reverse transcription of total RNA from FFPE tissue scrapes. The RNA quantity and quality is then measured using quantitative real-time PCR (Figure 1).

The RNA quantity and quality are assessed by the presence of intact RNA for a specific message. Two sets of β-actin gene-specific primers designed to amplify short segments of DNA within the β-actin target sequence are used to estimate the amount of RNA in a given sample in relation to β-actin content using the real-time PCR technique. The RNA quantity derived from the 3' primer set is used to quantitate the RNA in the FFPE tissue scrape. The ratio of the RNA yield obtained from both sets of PCR primers is the 3'⁄5' ratio and is used as an indication of RNA quality.

Key product features:
• Secure Downstream Success with the Paradise® QC Kit
• Benefit from the Only Complete System for Formalin-fixed Samples

Benefit from the Only Complete System for Formalin-fixed Samples
The Paradise® PLUS Reagent System is the only available solution that provides all the components necessary to prepare RNA from FFPE tissue samples for gene expression analysis. To ensure successful microarray results, the system includes reagents optimized for exceptional recovery of RNA and superior RNA amplification.

Integrated Systems for Microgenomics
The Paradise® PLUS Reagent System is validated as part of the ARCTURUS® Complete System for Microgenomics®, an integrated solution for utilizing small quantities of RNA for gene expression analysis, and features the ARCTURUSXT™ Laser Capture Microdissection (LCM) Instrument to procure pure cell populations. Arcturus' Systems for Microgenomics enable accurate and sensitive microarray assays that reveal molecular otherwise obscured in whole tissue samples or cell mixtures.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Downstream Application: Real-Time Quantitative PCR (qPCR)
For Use With (Equipment): ArcturusXT™ LCM Instrument
Product Line: ARCTURUS™, Paradise®
Product Size: 48 samples
Sample Type (General): FFPE & Fixed Samples
Sample Type (Specific): Tissue (FFPE)
Workflow Step: RNA Amplification, RNA Extraction, Reverse Transcription, Sample QC

Contents & storage

RNA Isolation kit (RNA extraction and purification reagents, purification columns), cDNA synthesis kit. Variable storage conditions.

Documents

Manuals & protocols