Applied Biosystems™

Arcturus™ Paradise™ PLUS 2 Round Kit

Catalog number:  KIT0322A

 Related applications: Laser Capture Microdissection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The ARCTURUS® Paradise® PLUS FFPE RNA Amplification Kit enables unprecedented gene expression analysis of millions of previously inaccessible formalin-fixed, paraffin-embedded (FFPE) tissue samples. Amplify RNA from as little as 5 ng of fixed RNA using reagents and methods that have been specially optimized to overcome the challenges of cross-linked FFPE templates (Figure 4).

Key product features:
• Integrate Seamlessly into Current Workflow
• Secure Downstream Success with the Paradise® QC Kit

Integrate Seamlessly into Current Workflow
The Paradise® PLUS Reagent System is compatible with all major microarray platforms. Robust and reproducible results are achieved using tools and techniques commonly employed in today’s research laboratories (Figures 1, 2 & 3).

Secure Downstream Success with the Paradise® QC Kit
Not all FFPE samples contain high quality RNA. Identify the best samples for your studies using a simple, real-time PCR research technique, and then proceed with confidence using these pre-qualified samples.

Integrated Systems for Microgenomics
The Paradise® PLUS Reagent System is validated as part of the ARCTURUS® Complete System for Microgenomics®, an integrated solution for utilizing small quantities of RNA for gene expression analysis, and features the ARCTURUSXT™ Laser Capture Microdissection (LCM) Instrument to procure pure cell populations. ARCTURUS®' Systems for Microgenomics enable accurate and sensitive microarray assays that reveal molecular otherwise obscured in whole tissue samples or cell mixtures.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Downstream Application: Microarray Analysis, Real-Time Quantitative PCR (qPCR)
For Use With (Equipment): ArcturusXT™ LCM Instrument
Includes Label or Dye: No
Product Line: ARCTURUS™, Paradise®
Product Size: 1 kit
Reverse Transcriptase: Not Included
Sample Type (General): FFPE & Fixed Samples
Sample Type (Specific): Tissue (FFPE)
Workflow Step: In Vitro Transcription, RNA Amplification, Reverse Transcription

Contents & storage

RNA Amplification Kit: cDNA synthesis reagents, in Vitro (IVT) reagents, amplification purification kit (purification reagents and columns). Variable storage conditions. Sufficient for 6 amplifications only.

Documents

Manuals & protocols