Histogene® Staining Solution

Catalog number: KIT0415

Applied Biosystems™  Related applications: Laser Capture Microdissection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This staining solution is for use with the ARCTURURS® HistoGene® LCM Frozen Section Staining Kit.

Designed as a fast-penetrating stain, it provides excellent contrast while preserving nucleic acid integrity and retaining low-abundance mRNAs in frozen tissue sections intended for laser capture microdissection (LCM).

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Downstream Application: Laser Capture Microdissection (LCM)
For Use With (Equipment): ArcturusXT™ LCM Instrument
Product Line: Applied Biosystems™, Histogene®
Product Size: 8 mL
Sample Type (General): Cells, Tissue
Sample Type (Specific): Tissue (Frozen)
Workflow Step: Tissue Staining

Contents & storage

Histogene® Staining Solution. Store at room temperature

Documents

Manuals & protocols