Applied Biosystems™

Arcturus™ RiboAmp™ HS PLUS cDNA Kit

Catalog number:  KIT0529

 Related applications: Laser Capture Microdissection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The ARCTURUS® RiboAmp® HS PLUS cDNA Kit permits quantitative Real-Time PCR (qRT-PCR) from as little as 100pg of total RNA. The kit uses random primers to enable robust reverse transcription across a broader transcript length.

Integrated Systems for Microgenomics
The ARCTURUS® Complete System for Microgenomics® offers an integrated solution for preparing small quantities of RNA for gene expression analysis. The System features the ARCTURUSXT™ Laser Capture Microdissection (LCM) Instrument to procure pure cell populations, the PicoPure® RNA Isolation Kit to extract and purify RNA, and the RiboAmp® PLUS Kit for reverse transcription and linear amplification of RNA. ARCTURUS® Systems for Microgenomics enable accurate and sensitive qRT-PCR and microarray assays that reveal molecular otherwise obscured in whole tissue samples or cell mixtures.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reverse Transcriptase: Not Included
Downstream Application: Real-Time Quantitative PCR (qPCR)
Reaction Format: SuperMix or Master Mix
Sample Type (General): Cells, Tissue
Final Product: cDNA (Double-Stranded)
Final Product Size(s): 2 kb or less
Optimal Reaction Temperature: 42° C
Product Line: ARCTURUS™, RiboAmp®
Product Size: 12 samples
Sample Type (Specific): Any Tissue Type, Cell Cultures, LCM Samples

Contents & storage

cDNA synthesis kit. Variable storage conditions

Documents

Manuals & protocols