Notice: This product will be discontinued when stock has been depleted. Catalog # BMS6002 is available as a replacement.
Invitrogen™

IL-1 beta Mouse ELISA Kit

Catalog number:  KMC0011

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The IL-1β Mouse ELISA Kit quantifies natural and recombinant mouse IL-1β in serum, plasma, buffered solution, or cell culture medium. IL-1β is important in immune response.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Cytokines & Receptors
Assay Incubation Time: 2.75 hr
Assay Range: 15.6-1000 pg/mL
Assay Sensitivity: <7 pg/mL
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Former Identifiers: Novex
Gene Aliases: IL-1beta, Il-1b
Gene ID: 16176
Gene Name: interleukin 1 beta
Gene Symbol: IL1B
Immunoassay Kit Format: Coated ELISA Kit
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 96 tests
Protein Aliases: IL-1 beta
Protein Name: Interleukin-1 beta
Sample Type (Specific): Cell Culture Supernatants, Plasma, Serum
Sample Volume: 50 µL
Shipping Condition: Wet Ice
Target (Kit Named): IL-1 beta
Target Species (Named): Mouse
Target Species (Validated): Mouse
UniProt ID: P10749

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols