IL-17A Mouse ELISA Kit

Catalog number: KMC3021

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Il-17 Mouse ELISA Kit is designed to quantify protein levels of mouse IL-17 in serum, plasma, and supernatant. Interleukin-17 (IL-17) plays an important role in bridging innate and adaptive immunity. This is an alternate product for Cat. No. KMC3022.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Cytokines & Receptors
Assay Incubation Time: 4 hr
Assay Range: 7.1-450 pg/mL
Assay Sensitivity: <5 pg/mL
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Former Identifiers: Novex
Gene Aliases: Ctla-8, Ctla8, IL-17, IL-17A, Il17
Gene ID: 16171
Gene Name: interleukin 17A
Gene Symbol: IL17A
Immunoassay Kit Format: Coated ELISA Kit
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 96 tests
Protein Aliases: IL-17, IL-17A, Cytotoxic T-lymphocyte-associated antigen 8, CTLA-8
Protein Name: Interleukin-17A
Sample Type (Specific): Cell Culture Supernatants, Plasma, Serum
Sample Volume: 100 µL
Shipping Condition: Wet Ice
Target (Kit Named): IL-17A
Target Species (Named): Mouse
Target Species (Validated): Mouse
UniProt ID: Q62386

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols