IL-2 ELISA Kit, Monkey

Catalog number: KPC0021

Novex™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The IL-2 Monkey ELISA Kit quantifies natural and recombinant monkey IL-2 in serum, buffered solution, or cell culture medium. IL-2 promotes T-cell growth.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gene Aliases: IL-2, TCGF, lymphokine
Accession Number: NP_000577.2, NM_000586.3
Shipping Condition: Wet Ice
Marker: IL-2
Gene ID: 3558
Species: Monkey
Gene Symbol: IL2
Sensitivity: <2 pg/mL
Target Gene: interleukin 2
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 96 assays
Concentration: 3.9-250 pg⁄ml
Sample Volume: 50 µl
Protein Family: Cytokines & Receptors
Incubation Time: 4 hrs
Detection Method: Colorimetric
Research Category: Immunology
Sample Type (Specific): Supernatants, Serum
For Use With (Equipment): Microplate Reader

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols