IL-2 Rat ELISA Kit

Catalog number: KRC0021

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The IL-2 Rat ELISA Kit quantifies natural and recombinant rat IL-2 in serum, buffered solution, or cell culture medium. IL-2 promotes T-cell growth.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Incubation Time: 3 hr
Assay Range: 23.4-1500 pg/mL
Assay Sensitivity: <5 pg/mL
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Former Identifiers: Novex
Gene ID: 116562
Gene Name: interleukin 2
Gene Symbol: IL2
Immunoassay Kit Format: Coated ELISA Kit
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 96 tests
Protein Aliases: IL-2, T-cell growth factor, TCGF
Protein Name: Interleukin-2
Sample Type (Specific): Cell Culture Supernatants, Serum
Sample Volume: 50 µL
Shipping Condition: Wet Ice
Target (Kit Named): IL-2
Target Species (Named): Rat
Target Species (Validated): Rat
UniProt ID: P17108

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols