xMAP® Classification Control Microspheres (CON1)

Catalog number: L100CON1

Novex™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

xMAP® Classification Control Microspheres (CON1), are polystyrene microspheres that are internally labeled with fluorescent dyes. These Control Microspheres are intended to verify the calibration and optical integrity for the Luminex 200™ System. Classification Control Microspheres verify both classification channels and the doublet discriminator channel.
The Luminex® 200™ System is sold separately.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Size: 5 mL
Channel Classification: CL2, Doublet Discriminator Channel (DD), CL1
For Use With (Equipment): Luminex® Instruments, Luminex® 200™ System
Shipping Condition: Wet Ice

Contents & storage

Store at 2-8°C.

Documents

Manuals & protocols

Recommended products