SuperScript™ Indirect cDNA Labeling Module, no purification columns

Catalog number:  L101403

Invitrogen™  Related applications: Nucleic Acid Labeling & Oligo Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The SuperScript® Indirect cDNA Labeling System is an array labeling kit based on proven methods of indirect cDNA labeling. The optimized system provides:

• SuperScript® III RT for generating high cDNA yields
• A proprietary nucleotide mixture to increase signal intensity
• A convenient kit format that saves valuable time

In this indirect labeling method, mRNA or total RNA is reverse transcribed using SuperScript® III RT, incorporating amino-modified dUTP and dATP into the synthesized cDNA. The template RNA is then degraded by base hydrolysis, and the reaction is neutralized with acid. The amino-modified cDNA is then purified to remove unincorporated nucleotides, primers, and buffers. In the second step, the modified cDNA is coupled with the active form of a fluorescent dye. The fluorescently labeled cDNA is purified with a S.N.A.P.™ spin column to remove any unreacted dye. The resulting fluorescently labeled cDNA is ready for hybridization to microarrays (Figure 1).

The SuperScript® Indirect cDNA Labeling System provides a complete system that includes both the core kit reagents (except dyes) and a purification module. The SuperScript® Indirect cDNA Labeling Module includes just the core kit reagents. Both provide the flexibility to use fluorescent nucleotides of your choice.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

LoyaltyEligible: SKUEligibleForLoyalty
Detection Method: Fluorescent
Final Product: cDNA (Labeled)
Includes Label or Dye: No
Label or Dye: Alexa Fluor™ 555, Alexa Fluor™ 647, Cy™3, Cy™5
Labeling Method: Indirect Labeling
Labeling Target: cDNA
Product Line: SuperScript™
Product Size: 30 reactions
Sample Type (General): RNA

Contents & storage

The SuperScript® Indirect cDNA Labeling Module includes SuperScript® III Reverse Transcriptase, oligo(dT)20- VN (2.5 µg/µl), random primers, control RNA ladder (0.5 g/µl), 5X first-strand buffer, 0.1 M DTT, 10 mM dNTP mixture, RNaseOUT™ Recombinant Ribonuclease Inhibitor (40 U/µl), DEPC-treated water, DMSO (Labeling Grade), 2X coupling buffer, 3M NaAc, pH 5.2, and glycogen (20 mg/ml). The SuperScript® Indirect cDNA Labeling System includes all of the components listed for the Labeling Module, plus S.N.A.P.™ column, collection tubes, loading buffer, wash buffer, amber collection tubes. Store all Labeling Module components at -20°C. Store S.N.A.P.™ columns, collection tubes, loading buffer, wash buffer, and amber collection tubes at room temperature. Guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products