Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

Low Density Lipoprotein from Human Plasma, Acetylated, Alexa Fluor™ 594 Conjugate (Alexa Fluor™ 594 AcLDL)

Catalog number: L35353

Invitrogen™  Related applications: Cell Viability, Proliferation & Function | Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Human low-density lipoprotein (LDL) is large protein complex (~500,000 Da) that binds to a specific receptor on the surface of vertebrate cells and delivers cholesterol via receptor-mediated endocytosis—our labeled LDL complexes are useful tools for studying this phenomenon. These experiments are typically performed by adding fluorescently labeled LDL to cultured cells and analyzing them by microscopy or flow cytometry. Alternatively the fluorescently labeled LDL can be injected into test animals, and the distribution of the label can be analyzed after the specified time period. We offer an unlabeled LDL and two classes of labeled LDLs: those containing an unmodified apoprotein (used to study normal cholesterol delivery and internalization) and those with an acetylated (Ac) apoprotein (used to study cell types that express receptors specific for this acetylated version (i.e., endothelial and microglial cells)).

LDL Specifications:

Label (Ex/Em): Alexa Fluor® 594 (590/617)
Acetylated: Yes
Amount: 200 µL (1.0 mg/mL)

Fresh LDL Produced Continually
We prepare our LDL and AcLDL products from fresh human plasma approximately every two months. The nonacetylated LDL products are shipped within two weeks of their preparation. All acetylated LDL products are available on a continuous basis.

Nonacetylated vs. Acetylated LDL
LDL containing an unmodified apoprotein is used to study normal cholesterol delivery and internalization. If the lysine residues of LDL’s apoprotein have been acetylated, the LDL complex no longer binds to the LDL receptor, but rather is taken up by endothelial and microglial cells that possess “scavenger” receptors specific for that modified form.

Key Applications for Labeled LDL
Some of the many applications for labeled LDL complexes include:
• Counting cell-surface LDL receptors, and analyzing their motion and clustering following internalization
• Quantitating LDL receptor activity in fibroblasts (replacing the radiolabeled LDL assay)
• Investigating LDL expression and identifying LDL receptor deficiencies in various cell lines

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: Alexa Fluor™
Product Size: 200 µL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products