Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells
Invitrogen™

Lucifer Yellow CH, Lithium Salt, 25 mg

Catalog number:  L453

 Related applications: Cell Tracing & Tracking | Cell Viability, Proliferation & Function

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Lucifer yellow CH, lithium salt is a water-soluble dye with excitation/emission peaks of 428/536 nm. It is a favorite tool for studying neuronal morphology, because it contains a carbohydrazide (CH) group that allows it to be covalently linked to surrounding biomolecules during aldehyde fixation. The lithium salt form of the lucifer yellow CH is commonly used for microinjection because it has higher solubility than the potassium and ammonium salt forms of lucifer yellow CH.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Yellow
For Use With (Equipment): Fluorescence Microscope
Sub-Cellular Localization: Cytoplasm & Cytosol
Detection Method: Fluorescent
Product Size: 25 mg
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature and protect from light.

Documents

Manuals & protocols