E-PAGE™ Blotting Pads (reusable), 8.6 cm x 13.5 cm

Catalog number: LC2101

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Invitrogen makes blotting of E-PAGE™ Gels easier and more efficient by providing a pre-cut fiber blotting pad and several types of pre-cut, pre-assembled membrane/filter paper sandwiches that perfectly fit 8.6 cm x 13.5 cm E-PAGE™ Gels*. The available nitrocellulose and Invitrolon™ PVDF membranes are designed for western transfer of proteins (Figure 1, Table 1). Pre-cut, extra-thick, absorbent Blotting Paper (2.5 mm thickness) for E-PAGE" Gels are also available to accommodate the E-PAGE™ blotting set-up using a semi-dry transfer apparatus. A protein s properties affect its ability to bind to membrane surfaces. Finding the right membrane may require experimenting with your specific protein on different membranes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gel Compatibility: E-PAGE™
Product Line: E-PAGE™, Novex™
Product Size: 4 pads
Type: Gel Blotting Accessories
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products