Invitrogen™

Novex™ Zymogram Developing Buffer (10X)

Catalog number: LC2671
Invitrogen™

Novex™ Zymogram Developing Buffer (10X)

Catalog number: LC2671
Catalog Number
LC2671
Unit Size
500 mL
Price (USD)
Price: 66.50
Your price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LC2671500 mL
Price: 66.50
Your price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Novex™ Zymogram Gels are excellent tools for detecting and characterizing proteases that utilize casein or gelatin as a substrate. Proteases are run on a gel containing one of these substrates under denaturing conditions and visualized as clear bands against a dark background following a simple renaturing, developing, and staining protocol. Zymogram gels are commonly used to detect matrix metalloproteinases.

Formulation: All Novex™ Zymogram Gels are Tris-Glycine gels with a substrate (gelatin or casein) incorporated into the gel. They are made with high-purity, strictly quality-controlled reagents: Tris base, HCl, acrylamide, bisacrylamide, TEMED, APS, highly purified water and casein or gelatin as the substrate. All have a 4% stacking gel.

Zymogram Gel Types: The 10% Zymogram (Gelatin) Gel is a 10% Tris-Glycine gel with 0.1% gelatin as the substrate. The 12% Novex™ Zymogram (Casein) Gel is a 12% Tris-Glycine gel with 0.05% casein as the substrate. The 4-16% Novex™ Zymogram (Blue Casein) Gel is a 4-16% Novex™ Tris-Glycine gel with 0.1% casein as the substrate and a proprietary dye incorporated throughout the gel.

Sensitivity Level:
10% Zymogram Gelatin Gel: 5 x 10-6 units of collagenase
12% Zymogram Casein Gel: 7 x 10-4 units of trypsin
4-16% Zymogram Casein Gel: 1.5 x 10-3 units of trypsin

Recommended Buffers: Use Novex™ Tris-Glycine SDS Sample Buffer and Novex™ Tris-Glycine SDS Running Buffer with these gels. Novex™ Zymogram Renaturing and Developing Buffers are also recommended.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer
Developing Buffers
Concentration
10 X
Gel Type
Zymogram
Product Type
Zymogram Developing Buffer
Gel Compatibility
Novex™ Zymogram Gels
Product Line
Novex™
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text