Novex™ Tris-Glycine Native Running Buffer (10X)

Catalog number:  LC2672

Invitrogen™  Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


Novex™ Tris-Glycine Native Running Buffer (10X) is designed for protein gel electrophoresis under native running conditions with NuPAGE™ Tris-Acetate gels or Novex™ Tris-Glycine gels.
For Research Use Only. Not for use in diagnostic procedures.


Gel Type: Native Gel
Shipping Condition: Room Temperature
Buffer Type: Running Buffers
Concentrated: 10 X
Gel Compatibility: Novex® Tris-Glycine Gels, NuPAGE® Tris-Acetate Gels
Product Line: Novex™
Product Size: 500 mL

Contents & storage

Store at room temperature


Manuals & protocols

Recommended products