Novex™ Tris-Glycine SDS Running Buffer (10X)

Catalog number:  LC26754

Invitrogen™  Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Novex® Tris-Glycine SDS Running Buffer (10X) is formulated for separation of proteins, in their denatured state, on Novex® Tris-Glycine gels. Tris-Glycine gels provide reproducible separation of a wide range of proteins into well-resolved bands, depending upon the gel percentage used.

Separate native or denatured proteins
Novex®Tris-Glycine gels do not contain SDS and can be used to accurately separate both native and denatured proteins, depending upon the sample and running buffers used. To separate denatured proteins on Novex® Tris-Glycine gels, use Novex® Tris-Glycine SDS Sample Buffer and Novex® Tris-Glycine SDS Running Buffer. To separate native proteins on Novex® Tris-Glycine gels, use Novex® Tris-Glycine Native Sample Buffer and Novex® Tris-Glycine Native Running Buffer.

Easy-to-use format
Pre-mixed buffers are a convenient way to ensure high-quality, consistent electrophoresis results. All buffers are made with high-purity reagents and are strictly quality controlled. The buffers are provided as a concentrated solution requiring a simple dilution with deionized water before use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gel Type: SDS PAGE Gel
LoyaltyEligible: SKUEligibleForLoyalty
Shipping Condition: Room Temperature
Buffer Type: Running Buffers
Concentrated: 10 X
Gel Compatibility: Novex® Tris-Glycine Gels
Product Line: Novex™
Product Size: 4 x 1 L

Contents & storage

Includes Novex® Tris-Glycine SDS Running Buffer (10X denaturing, 4 × 1 L)

Documents

Manuals & protocols

Recommended products