Novex™ IEF Cathode Buffer pH 3-10 (10X)

Catalog number:  LC5310

Invitrogen™  Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Novex® IEF Cathode Buffer pH 3-10 (10X) is optimized pre-mixed IEF cathod buffer for using with Novex® pH 3-10 IEF gels. IEF is a sensitive technique which is affected by many factors. The optimized pre-mixed IEF buffers reduce variability and ensure consistent results.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gel Type: IEF Gel
Shipping Condition: Wet Ice
Buffer Type: Anode & Cathode Buffers
Concentrated: 10 X
Gel Compatibility: Novex® IEF Gels
Product Line: Novex™
Product Size: 125 mL

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products